BLOCK-iT™ RNAi Express
READY, CLICK and GO: Fast and easy online ordering of bench-tested Validated Stealth RNAi™ siRNA, ready-to-order Stealth Select RNAi™ siRNA, bench-tested BLOCK-iT™ Pol II miR Validated miRNA Vector Duopaks.
 RNAi Express Options:  Search By ID/Keyword Browse By Pathways Browse By Gene Families
TGF-β/BMP Pathway
The TGF-β superfamily regulates a variety of cellular processes including proliferation, differentiation, and apoptosis. Additionally, members of the TGF-β family are also key regulators of embryonic development and tissue differentiation.
TGF-β, bone morphogenic proteins (BMPs), and activins function by activation of a transmembrane serine-threonine kinase receptor composed of type I and type II subunits. In response to ligand binding, the type II subunit phosphorylates and functionally activates the type I subunit. The activated type I subunit subsequently phosphorylates receptor-regulated Smad proteins (R-Smad: Smad1, Smad2, Smad 3, Smad5, and Smad8) on C-terminal serine residues. These activated R-Smad proteins form a heterodimer with Smad4, translocate to the nucleus, and regulate gene expression. I-Smad proteins (Smad6 and Smad7) negatively regulate TGF-β signaling by competing with R-Smads for type I receptor binding sites and recruitment of E3-ubiquitin ligases (Smurf 1 and Smurf 2).
TGF-β/BMP Pathway Validated Stealth RNAi™ siRNA
TGF-β/BMP Receptors
SMAD
Smad ubiquitin regulatory factors (SMURF)
References
1. Attisano, L & Wrana, J.L. Signal transduction by the TGF-b superfamily. Science 296, 1646-1647 (2002).
2. Derynck, R. & Zhang, Y.E. Smad-dependent and Smad-independent pathways in TGF-b family signaling. Nature 425, 577-584 (2003).
3. Dijke, P. & Hill. C.S. New insights into TGF-b-Smad signaling. Trends Biochem. Sci. 29, 265-273 (2004).
4. Shi, Y. & Massague, J. Mechanisms of TGF-b?signaling from cell membrane to the nucleus. Cell 113, 685-700 (2003).